missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

Catalog No. AA1321409
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1321409 10 g
AA1321404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1321409 Supplier Thermo Scientific Chemicals Supplier No. 01321409

CAS: 14553-08-3 | C15H21ErO6 | 464.59 g/mol

Erbium(III) 2,4-pentanedionate is commonly used in various catalysts and catalytic reagents for organic synthesis. It is also used for glass coloring, as an amplifier in fiber optics, and in lasers for medical and dental purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14553-08-3
Molecular Formula C15H21ErO6
Molecular Weight (g/mol) 464.59
MDL Number MFCD00013490
InChI Key FGQSJRDKBCVFHH-UHFFFAOYSA-N
Synonym erbium 2,4-pentanedionate, erbium iii acetylacetonate hydrate, erbium iii 2,4-pentanedionate, reacton, 3z-4-bis 2z-4-oxopent-2-en-2-yl oxy erbio oxy pent-3-en-2-one
PubChem CID 102601666
SMILES [Er+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
IUPAC Name erbium;(Z)-4-hydroxypent-3-en-2-one
Formula Weight 464.59
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Erbium(III) 2, 4-pentanedionate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only