missing translation for 'onlineSavingsMsg'
Learn More

Erbium foil, 0.25mm (0.01in) thick, 99.9% (REO)

Catalog No. p-7048989
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12386FF 25 x 25 mm
AA12386FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12386FF Supplier Thermo Scientific Chemicals Supplier No. 012386FF
Only null left

CAS: 7440-52-0 | Er | 167.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Weight ≈1.4g/25x25mm
Form Foil
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Erbium foil, 0.25mm (0.01 in.) thick
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
EINECSNumber 231-160-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only