Learn More
United Scientific™ Equilateral Refraction Prism
Catalog No. S15984
Change view
Click to view available options
Material:
Acrylic
Flint Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S15984 Flint Glass
S15807 Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15984 Supplier United Scientific™ Supplier No. FGP075
Ideal for classroom use.
  • Acrylic, or Flint glass prism, ideal for classroom use.
  • Measures 75 mm long x 9 mm thick
  • Clear optical properties for experiments
Specifications
Material Flint Glass
Product Type Prism
For Use With (Application) Optics and Light