Learn More
Eisco™ Equilateral Prisms
Catalog No. S41533
Change view
Click to view available options
Length (Metric):
25 mm
50 mm
75 mm
100 mm
150 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S41533 150 mm
S41529 25 mm
S41530 50 mm
S41531 75 mm
S41532 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S41533 Supplier Eisco™ Supplier No. PH0554LG
These equilateral prisms are polished on all faces. Designed for use in optical experiments. Prisms will create a nice rainbow.
  • Made with optical quality glass or clear acrylic
  • Polished on all faces
  • Designed for use in optical experiments
  • Will create a nice rainbow
Warranty
1 year limited manufacturer warranty
Specifications
Product Type Equilateral Solid
Material Glass
For Use With (Application) Optics experiments and demonstrations
Quantity 1
Length (Metric) 150 mm
Weight (Metric) 160 g