missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epT.I.P.S.™ Box 2.0 - Pipette Tips
GREENER_CHOICE

Catalog No. 05414186
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
0.1 to 5.0 mL
0.25 to 2.5 mL
0.5 to 20 μL
2 to 200 μL
20 to 300 μL
50 to 1000 μL
50 to 1250 μL
Color Code:
Blue
Dark Gray
Dark Green
Green
Light Gray
Medium Gray
Orange
Red
Violet
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Code
05414186 50 to 1000 μL Blue
05414181 0.1 to 10 μL Dark Gray
05414182 0.1 to 20 μL Medium Gray
05414183 0.5 to 20 μL Light Gray
05414184 2 to 200 μL Yellow
05414185 20 to 300 μL Orange
05414187 50 to 1250 μL Green
05414188 50 to 1250 μL Dark Green
05414189 0.25 to 2.5 mL Red
05414190 0.1 to 5.0 mL Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 05414186 Supplier Eppendorf™ Supplier No. 0030076176
Only null left

Eppendorf epT.I.P.S.™ Box 2.0, available with 'Eppendorf Quality' pipette tips or as an empty box, improves lab efficiency with innovative TwinLid™ technology, enhanced stackability, and superior ergonomics for safe, organized pipetting.

  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121°C for subsequent use
  • epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability

Specifications

Volume (Metric) 50 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Model epTIPS
Clean Claims Eppendorf Quality (Non-sterile)
Format Box
Packaging 1 Reusable Box x 96 Tips
Quantity 96 Tips/Cs.
Product Type Pipette Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Air-Cushion System
Graduated Yes
Product Line epT.I.P.S.
Material Polypropylene
Show More Show Less