missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ epT.I.P.S.™ 384 Reloads - Micropipette Tips

Description
- epT.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16- and 24-channel pipettes as well as 20 μL and 100 μL Move It adjustable tip spacing pipettes for utmost safety and reliability
- Designed for a cone distance of 4.5 mm
- Innovative SOFTattach technology allows for reliable tip fit and tightness
- Fill a whole 384-well plate without loosening pipette tips
- Perfect tip alignment by extraordinary coaxiality for safe plate maneuvering and liquid transfer
- The tip shape of the epT.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
- Tip attachment forces reduced by 40% per cone compared to 8- and 12-channel pipettes
- Tip ejection forces in 16- and 24-channel pipettes reduced by SOFTeject for ergonomic working conditions
- Available as reload, and set
- Available in purity grades of Eppendorf Quality and PCR clean
- ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
Specifications
Specifications
| Volume (Metric) | 5 to 100 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Colorless |
| Autoclavable | Autoclavable |
| Model | epT.I.P.S.™ 384 Reloads |
| Length (English) | 2.1 in. |
| Length (Metric) | 53 mm |
| Clean Claims | PCR Clean |
| Storage Requirements | Room temperature |
| Show More |
The pipette tips of the :Productname: product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned :Productname: are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.