missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epT.I.P.S.™ 384 Reloads - Micropipette Tips
GREENER_CHOICE

Catalog No. 05413607
Change view
Click to view available options
Volume (Metric):
0.1 to 20 μL
5 to 100 μL
Clean Claims:
Eppendorf Quality
PCR Clean
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Clean Claims
05413607 0.1 to 20 μL PCR Clean
05413610 5 to 100 μL PCR Clean
05413608 0.1 to 20 μL Eppendorf Quality
05413611 5 to 100 μL Eppendorf Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05413607 Supplier Eppendorf™ Supplier No. 0030076001
Only null left

Eppendorf epT.I.P.S.™ 384 Reloads provide high-precision micropipette tips in 'Eppendorf Quality' and 'PCR clean' grades, tailored for 384-well plate applications, ensuring accuracy, reduced waste, and compatibility in high-throughput labs.

  • epT.I.P.S. 384 pipette tips build a system together with the Research plus and Xplorer plus 16- and 24-channel pipettes as well as 20 μL and 100 μL Move It adjustable tip spacing pipettes for utmost safety and reliability
  • Designed for a cone distance of 4.5 mm
  • Innovative SOFTattach technology allows for reliable tip fit and tightness
  • Fill a whole 384-well plate without loosening pipette tips
  • Perfect tip alignment by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of the epT.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Tip attachment forces reduced by 40% per cone compared to 8- and 12-channel pipettes
  • Tip ejection forces in 16- and 24-channel pipettes reduced by SOFTeject for ergonomic working conditions
  • Available as reload, and set
  • Available in purity grades of Eppendorf Quality and PCR clean
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability

Specifications

Volume (Metric) 0.1 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Model epT.I.P.S.™ 384 Reloads
Length (English) 1.7 in.
Length (Metric) 42 mm
Clean Claims PCR Clean
Storage Requirements Room temperature
Format Refill
Tip Style Universal
Packaging 3,840 Tips (10 Trays x 384 Tips)
Quantity 3840 Tips
Product Type Micropipette Tips
Color Code Light Pink
Disposable Single-use
For Use With (Equipment) Air-Cushion System, Universal
Product Line epT.I.P.S.
Material Polypropylene
Show More Show Less

The pipette tips of the :Productname: product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned :Productname: are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.