missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactone 99.0+%, TCI America™

Catalog No. C070225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C070225ML 25 mL
C0702500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C070225ML Supplier TCI America Supplier No. C070225ML

Chemical Identifiers

CAS 502-44-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00003267
InChI Key PAPBSGBWRJIAAV-UHFFFAOYSA-N
Synonym 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
PubChem CID 10401
ChEBI CHEBI:17915
IUPAC Name oxepan-2-one
SMILES C1CCC(=O)OCC1

Specifications

Melting Point -1°C
Color Yellow
Boiling Point 140°C
Quantity 25 mL
Formula Weight 114.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material epsilon-Caprolactone
EINECSNumber (5)-1091
RTECSNumber MO8400000
TSCA Yes