Learn More
Eppendorf™ Vis Cuvettes
Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the ideal tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.
Supplier:  Eppendorf™ 0030079345
 View more versions of this product
Catalog No. 14-281-216
Only null left
Description
Description
  • Two different cuvette sizes (semi-micro and macro) for a wide variety of volumes and applications
  • Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm
Specifications
Specifications
Macro Vis Cuvette
Plastic
Product Suggestions
Product Suggestions