Learn More
Eppendorf™ Vis Cuvettes
Catalog No. 14281215
Change view
Click to view available options
Type:
Macro Vis Cuvette
Semi-micro Vis Cuvette
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14281215 Semi-micro Vis Cuvette
14281216 Macro Vis Cuvette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14281215 Supplier Eppendorf™ Supplier No. 0030079353
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the ideal tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.
  • Two different cuvette sizes (semi-micro and macro) for a wide variety of volumes and applications
  • Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm
Specifications
Type Semi-micro Vis Cuvette
Material Plastic