missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates 384-well, 45 μL - PCR Plates

Catalog No. 361023837
Change view
Click to view available options
Color:
Blue
Colorless
Green
Red
Yellow
Barcode:
Alphanumeric
No Barcode
Packaging:
10 Bags x 30 Plates
5 Bags x 5 Plates
Individually Blister Wrapped
Purity or Quality Grade:
PCR Clean (Non-sterile)
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Barcode Packaging Purity or Quality Grade
361023837 Colorless No Barcode Individually Blister Wrapped PCR Clean (Non-sterile)
361023838 Blue No Barcode Individually Blister Wrapped PCR Clean (Non-sterile)
E951020737 Blue No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020711 Yellow No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020729 Green No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020745 Red No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020702 Colorless No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
14380055 Colorless No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
361030806 Colorless No Barcode 10 Bags x 30 Plates PCR Clean (Non-sterile)
361030811 Blue No Barcode 10 Bags x 30 Plates PCR Clean (Non-sterile)
01671102 Colorless Alphanumeric 5 Bags x 5 Plates PCR Clean (Non-sterile)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 361023837 Supplier Eppendorf™ Supplier No. 0030129342

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean).
  • Safe Code option provides 3-level coding for safe sample and vessel identification.
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes.

Certifications

Certified free of human DNA, DNase, RNase, and PCR inhibitors

Specifications

Barcode No Barcode
Certifications/Compliance Free of Human DNA, DNase, RNase and PCR Inhibitors
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Packaging Individually Blister Wrapped
Color Colorless
Product Type PCR Plate
For Use With (Equipment) Automated Systems
Skirt Style skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 45 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Round Bottom (U-well)
Max. RCF 2250 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 10 Pk.
Well Color Colorless
Show More Show Less