missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Tubes™ with Screw Cap 5.0 mL - Microtube
GREENER_CHOICE

Catalog No. 13864407
Change view
Click to view available options
Capacity (Metric):
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13864407 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13864407 Supplier Eppendorf™ Supplier No. 0030122321
Get More Eppendorf Consumables with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eppendorf Tubes™ with screw cap are your ideal choice when working with medium-sized sample volumes (0.5 - 5.0 mL). SafeCode variants enhance sample identification, while the BioBased option, manufactured using renewable resources, provides an eco-friendly alternative without compromising performance.

Use these tubes for the simple and safe processing of sample volumes up to 5.0mL. Eppendorf Tubes™ represent the perfect option for working with medium–sized sample volumes.

BioBased (made of bio-based material) variant aids in the reduction of consumable-related carbon footprint in the lab. The bio-based tubes have identical material properties and technical specifications as our non-BioBased tubes.

  • Ensures leak-free storage and protects sample integrity.
  • Suited for a wide range of laboratory tasks, enhancing flexibility.
  • High-quality materials for long-lasting, robust performance.
  • Clear, visible graduations for quick volume assessment.
  • Facilitates optimal sample recovery and efficient centrifugation.
  • Available in Eppendorf Quality, PCR Clean, Sterile/Pyrogen-/DNase-/RNase-/Human and Bacterial DNA-free, and Forensic DNA Grade quality for tested purity.
  • 2D Datamatrix (SafeCode) option features 2-level coding for safe sample identification.
  • ECC200 error correction function enables safe code reading with up to 25% damaged codes.

Specifications

Certifications/Compliance Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free
Closure Material HDPE
Color Colorless
Capacity (Metric) 5 mL
Material Polypropylene
For Use With Easy processing of samples up to 5.0 mL, DNA isolation, especially with samples of limited quantity of source material, Preparation of mastermixes and buffers, Cell and tissue lysis
Includes Microtubes
No. per Pack 100/Pk.
Packaging 200 Tubes (2 Bags x 100 Tubes)
Quantity 200 Pk.
Sterility Sterile
Autoclavable Not Autoclavable
Capacity (English) 0.169 oz.
Clean Claims Pyrogen, Dnase, Rnase, human and bacterial DNA free
Bottom Shape Conical
Closure Type Screw Cap
Temperature Range (Metric) -86°C to +100°C
Diameter (Metric) 17 mm
Graduated Yes
Product Type Microcentrifuge Tube
Max. RCF 25,000 x g
Purity or Quality Grade Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free
Show More Show Less