missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ ThermoStat™ C

Catalog No. 05412502
Change view
Click to view available options
Height (English):
5.2 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
05412502 5.2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05412502 Supplier Eppendorf™ Supplier No. 5383000027

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sets and maintains temperature at high accuracy for all conventional lab vessels

  • Peltier technology enables comfortable and precise temperature control from -10°C to +110°C
  • Heating and active cooling in all common vessel and plate formats from 5μL to 50mL
  • Precise maintenance of various temperature levels
  • Reliable prevention of condensate formation with optional ThermoTop™ (condens.protect™) lid
  • Highly flexible due to large selection of exchangeable SmartBlocks
  • Dry incubation for improved lab hygiene
  • Simple and intuitive operation due to predefined temperature keys (4, 16, 37, 56 and 95°C)

  • Individually store up to 15 programs
  • Comprehensive range of program functions
  • Quick release system on SmartBlock™ for 2 sec. block exchange

Specifications

Accuracy ±0.5°C at 20 to 45°C
Temperature (English) Maximum 230°F
Height (English) 5.2 in.
Height (Metric) 13.1 cm
Temperature (Metric) Maximum 110°C
Width (Metric) 20.6 cm
Wattage 200w
Width (English) 8.1 in.
Type Temperature Controller
For Use With (Application) Gentle thawing and cooling buffers and samples at low temperature, preheating media, biochemical reactions at low temperature, ligation, cDNA synthesis, denaturation of RNA, DNA and protein, Proteinase K digestion, ELISA assays, Immunoprecipitation, cell
Voltage 120 V
Show More Show Less