Learn More
Eppendorf™ ThermoTop™
Catalog No. 05412516
Change view
Click to view available options
Type:
Tube Lid
Catalog No. Type
05412516 Tube Lid
1 options
Catalog No. 05412516 Supplier Eppendorf™ Supplier No. 5308000003
Use this lid with ThermoMixer™ C/ ThermoStat™ C with SmartBlock™ plates and lab vessels up to 2 mL and ThermoMixer F 0.5/F1.5/F2.0/FP. The Eppendorf ThermoTop™ with condens.protect™ technology heated lid prevents condensation on tube lid and tube wall.
  • Efficient and reliable prevention of condensation on tube lid and wall
  • Get constant reaction conditions for optimal enzymatic performance within your samples
  • Wireless connection with automatic recognition and operation for user-friendly handling
Specifications
Capacity (Metric) 2 mL
For Use With (Equipment) Thermomixer F1.5, FP and C, ThermoStat C with Eppendorf SmartBlocks 0.5, 1.5 and 2mL, Plates, PCR 96 and PCR 384
Type Tube Lid
For Use With (Application) Plates, lab vessels up to 2 mL