Learn More
Eppendorf™ Storage Foil Adhesive Seal for Microplates
Storage Film and Foil glued seals for simple and dependable sealing during sample storage
Supplier:  Eppendorf™ 0030127889
Catalog No. E0030127889
Only null left
Explore available promotions
Description
Description
  • Heat Sealing Film and Foil for continuous locking with the best evaporation protection
  • Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
  • Film products are transparent and provide protection against unwanted punctures
  • Eppendorf Plate Lid: stable, flexible protection of samples during short-term storage
Specifications
Specifications
Aluminum
Storage Foil
Product Suggestions
Product Suggestions