Learn More
Eppendorf™ Eppendorf™ Aerosol-tight Caps
Catalog No. 5820747005
Change view
Click to view available options
For Use With (Equipment):
Microtitor plate with 750mL bucket in S-4-104 rotor with 5810/5810R centrifuge
Catalog No. For Use With (Equipment)
5820747005 Microtitor plate with 750mL bucket in S-4-104 rotor with 5810/5810R centrifuge
1 options
Catalog No. 5820747005 Supplier Eppendorf™ Supplier No. 5820747005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use these caps for 150mL round buckets. Eppendorf™ Aerosol-tight caps are for use with Rotor S-4-104, Rotor S-4x750 and Rotor S-4x1000.ids
Come in a pack of two
Specifications
For Use With (Equipment) Microtitor plate with 750mL bucket in S-4-104 rotor with 5810/5810R centrifuge