Learn More
Eppendorf™ S-4-104 Rotor Adapters
Available in a wide range of configurations to suit a variety of applications.
Supplier:  Eppendorf™ 5825745000
 View more versions of this product
Catalog No. 5825745000
Only null left
Explore available promotions
Includes: 2 piece set
Description
Description
  • For use with Centrifuge 5810/5810 R
  • Holds up to 4 × 750mL round buckets
Specifications
Specifications
Rotor Adapter
1 x 500 mL Bottle
Rotor S-4-104, Rotor S-4x1000 round buckets and Rotor S-4x750, set of 2 pcs
2 Pack
Product Suggestions
Product Suggestions