Learn More
Eppendorf™ S-4-104 Rotor Adapters
Available in a wide range of configurations to suit a variety of applications.
Supplier:  Eppendorf™ 5825733002
 View more versions of this product
Catalog No. 5825733002
Only null left
Explore available promotions
Includes: 2 piece set
Description
Description
  • For use with Centrifuge 5810/5810 R
Specifications
Specifications
Rotor Adapter
7 x 50 mL Conical Tubes
S-4-104 rotor for 5810/5810R centrifuge
2 Pack
Product Suggestions
Product Suggestions