missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ O-rings, red

Catalog No. EPP3122611000
Change view
Click to view available options
Quantity:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EPP3122611000 Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EPP3122611000 Supplier Eppendorf™ Supplier No. 3122611000

Chemically resistant. Red Eppendorf™ O-rings are available in packs of 24.

  • Compatible with 50, 100 and 300μL multi-channel pipettes

Specifications

For Use With (Equipment) Research multi 100ul, 300ul; Research Plus Multi 100ul, 300ul; Research Pro 100ul, 300ul; Newer style
Quantity Pack of 24
Product Type O-Ring
Color Red