missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ PCR Cooler

Catalog No. p-322983
Change view
Click to view available options
Color:
Blue
Pink
Set of 1 Pink and 1 Blue
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0540300 Blue
0540299 Pink
0540301 Set of 1 Pink and 1 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0540300 Supplier Eppendorf™ Supplier No. 022510525
Only null left

Maintains sample temperature for over an hour; changes color when temperature exceeds 7°C

  • Maintains sample temperature at 0°C
  • Temperature of each sample is indicated by the color of its individual well
  • Cone-shaped contours prevent samples from freezing
  • Accommodates 0.2mL PCR tubes (individual, tube strip, or 96-well plate)

Specifications

Color Blue
Volume (Metric) 0.2 mL