Learn More
Eppendorf™ Model 5424 Microcentrifuge Rotor Kit
Catalog No. 05400012
Change view
Click to view available options
For Use With (Equipment):
5430 Microcentrifuge
Catalog No. For Use With (Equipment)
05400012 5430 Microcentrifuge
1 options
Catalog No. 05400012 Supplier Eppendorf™ Supplier No. 022653083
Only null left
Includes
Rotor and lid
For use with Eppendorf Model 5424 Microcentrifuges
18-place Miniprep Spin Column Rotor Kit
  • Deep lip design supports open microtube lids
  • Capacity: 18 × 1.5/2.0mL tubes
  • Max.RCF: 18,111×G
  • Supplied with lid 05-400-013
Order Info
Replacement lids are available (Cat. No. 05-400-011).
Specifications
Type Rotor Kit
Material Aluminum, Polypropylene Lid
For Use With (Equipment) 5430 Microcentrifuge
Capacity 18 x 1.5/2 mL
Height (Metric) 22.7 cm
Includes Extra high rimmed lid