Learn More
Eppendorf™ Microcentrifuge 5430 Accessories: PCR Plate Adapters
Designed to support thin-walled PCR plate wells during centrifugation
$471.34
Specifications
For Use With (Equipment) MTP/DWP rotor bucket for 5810/5810R/5430 centrifuge
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
04987567
View Documents Promotion Details
Eppendorf™
022638955
348-well PCR Plate Adapter
Pair for $471.34
Only null left
04987566
View Documents Promotion Details
Eppendorf™
022638947
96-well PCR Plate Adapter
Pair for $471.34
Only null left
Description
Description
  • Available for 96-well and 384-well PCR plates
  • Adapters can be used in all Eppendorf plate rotors
Specifications
Specifications
MTP/DWP rotor bucket for 5810/5810R/5430 centrifuge