missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Maxitips - Pipette Tips

Catalog No. 2137951
Change view
Click to view available options
Volume (Metric):
2.5 to 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
2137951 2.5 to 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2137951 Supplier Eppendorf™ Supplier No. 022291301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eppendorf Maxitips are designed for large volume liquid handling, offering high precision and compatibility with Eppendorf pipetting systems. Ideal for diverse lab applications, they ensure accuracy and ease of use, enhancing workflow efficiency.

  • Use Maxitips P to aspirate 1 mL to 10 mL from beakers using the positive displacement principle
  • Use Maxitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL liquid from high, narrow-mouth vessels with the air cushion principle
  • Valve for Maxitips ensures drip-free dispensing of liquids with a high vapour pressure
  • Graduation on Maxitips ensures accurate dispensing of non-aqueous liquids

Specifications

Description Maxitip S Kit
Volume (Metric) 2.5 to 10 mL
Filtered Non-filtered
For Use With (Application) Aspirating 2.5 mL to 10 mL from tall, narrow-neck vessels using an air-displacement technique
Tip Style Positive Displacement, Macro
Quantity 100 Maxitips, 10 Dispensing Parts, 10 Valves
Product Type Pipette Tips
For Use With (Equipment) Positive Displacement System