missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ PCR Cap Strips, Domed
GREENER_CHOICE

Catalog No. E0030124839
Change view
Click to view available options
Quantity:
120 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0030124839 120 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0030124839 Supplier Eppendorf™ Supplier No. 0030124839

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

All the benefits of white reflective wells in a PCR-tube format

  • Strips with eight microcaps for 0.1 and 0.2 mL wells
  • Easy and rapid sealing of Eppendorf twin.tec PCR Plates, PCR strips and other standard multiwell plates
  • Autoclavable (121 °C, 20 min)
  • Certified free from human DNA, DNase, RNase and PCR inhibitors

Specifications

Certifications/Compliance Human DNA, DNase, RNase, PCR inhibitor-free
Quantity 120 Pk.
Closure Material Polypropylene
For Use With (Equipment) 0.1 and 0.2mL tube strips, PCR plates and other standard multiwell plates by providing easy and effective sealing
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Autoclavable Autoclavable
Format 10 x 12 Strips
Product Type 8-Strip Domed Caps (no tubes)