Learn More
Eppendorf™ Excella™ E-25 Console Biological Shaker Shaker with Platform and Clamps
Feedback control and fans maintain temperature when lid is open
Supplier:  Eppendorf™ M13530024
Catalog No. 14285895
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Microprocessor controls temperature and shaking speed
  • Available in choice of incubated and incubated/refrigerated shakers
  • 1 in. Orbit with 30 in.×18 in. Platform and Clamps Bundle
  • 30L × 18 in.W (762 × 457mm)
  • 120V 60Hz
Specifications
Specifications
30 x 18 in. (76.2 x 45.7 cm)
76.2 cm
Incubated/Refrigerated
30 in.
45.7 cm
Product Suggestions
Product Suggestions