missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epT.I.P.S.™ Reloads - Pipette Tips

Catalog No. 0540339
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
0.25 to 2.5 mL
0.5 to 20 μL
2 to 200 μL
20 to 300 μL
50 to 1000 μL
50 to 1250 μL
50 to 2500 μL
Clean Claims:
Eppendorf Quality
PCR Clean
Color Code:
Blue
Dark Gray
Dark Green
Green
Light Gray
Medium Gray
Orange
Red
Yellow
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Clean Claims Color Code
0540339 0.1 to 20 μL Eppendorf Quality Medium Gray
0540338 0.1 to 10 μL Eppendorf Quality Dark Gray
0540340 0.5 to 20 μL Eppendorf Quality Light Gray
0540341 2 to 200 μL Eppendorf Quality Yellow
0540342 20 to 300 μL Eppendorf Quality Orange
0540343 50 to 1000 μL Eppendorf Quality Blue
0540344 50 to 1250 μL Eppendorf Quality Green
05412301 50 to 1250 μL Eppendorf Quality Dark Green
0540345 0.25 to 2.5 mL Eppendorf Quality Red
0540346 0.1 to 10 μL PCR Clean Dark Gray
0540347 0.1 to 20 μL PCR Clean Medium Gray
0540348 0.5 to 20 μL PCR Clean Light Gray
0540349 2 to 200 μL PCR Clean Yellow
0540350 20 to 300 μL PCR Clean Orange
0540351 50 to 1000 μL PCR Clean Blue
0540352 50 to 1250 μL PCR Clean Green
05412302 50 to 1250 μL PCR Clean Dark Green
0540353 50 to 2500 μL PCR Clean Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 0540339 Supplier Eppendorf™ Supplier No. 022491512
Get More Eppendorf Consumables with Purchase
View all promotions
Edge

Eppendorf epT.I.P.S.™ Reloads offer high-precision pipette tips in 'Eppendorf Quality' and 'PCR clean' purity grades. Eco-friendly, cost-effective, and designed for seamless compatibility with Eppendorf Box 2.0, they ensure accuracy and sustainability in labs.

  • Reduced waste as compared with disposable racks
  • The refill trays can be autoclaved up to 121 °C, in packaging and without foil, for subsequent use
  • The refill system, depending on tip size, is packaged as either dual-sided or in stack form
  • Available in purity grades of Eppendorf Quality and PCR clean
  • Color-coded trays for simple volume and pipette identification
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability

Specifications

Volume (Metric) 0.1 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Length (English) 1.57 in.
Length (Metric) 40 mm
Clean Claims Eppendorf Quality
For Use With (Application) Reliable, reproducible, high-precision pipetting for standard laboratory use
Format Refill
Tip Style Extended Length, Finely Tapered, Universal
Packaging 960 Tips (10 Trays x 96 Tips)
Product Type Pipette Tips
Color Code Medium Gray
Disposable Single-use
For Use With (Equipment) Air-Cushion System
Graduated Yes
Product Line epT.I.P.S.
Material Polypropylene
Show More Show Less