missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Combitips™ Advanced, Sterile - Individually Wrapped Positive Displacement Pipette Tips

Catalog No. 13683717
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
Color Code:
Blue
Green
Light Blue
Light Gray
Orange
Red
Violet
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Code
13683717 50 mL Light Gray
13683709 0.1 mL White
13683710 0.2 mL Light Blue
13683711 0.5 mL Violet
13683712 1 mL Yellow
13683713 2.5 mL Green
13683714 5 mL Blue
13683715 10 mL Orange
13683716 25 mL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13683717 Supplier Eppendorf™ Supplier No. 0030089596
Edge

Eppendorf's consumable of choice for Eppendorf Repeater™ multi-dispensers. Available in 9 volume sizes (0.1 to 50 mL), Combitips advanced in 'Sterile Quality' dispense in increments as low as 100 mL and are ideally suited for a variety of standard lab applications. Also available in purity grades Eppendorf Quality, PCR clean, Biopur™ or Forensic DNA Grade.

  • Combitips advanced are the ideal system components for all Eppendorf Multipettes
  • The 9 volume sizes (0.1 mL – 50 mL) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters depending on the Combitip used
  • Elongated Combitips in volumes 2.5 mL, 5.0 mL and 10 mL make it possible to reach the bottom of the most common laboratory vessels
  • An individual color coding facilitates the quick identification of the desired Combitips

Specifications

Volume (Metric) 50 mL
Sterility Sterile
Clean Claims Sterile
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk., Individually wrapped
Product Type Pipette Tips
Color Code Light Gray
For Use With (Equipment) Eppendorf Multipette
Graduated Yes
Product Line CombiTips Advanced