missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - Eppendorf Quality
GREENER_CHOICE

Catalog No. p-4909476
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1.0 mL
2.5 mL
5.0 mL
10 mL
25 mL
50 mL
Color Code:
Blue
Green
Light Blue
Light Gray
Orange
Red
Violet
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Code
13683700 0.1 mL White
13683701 0.2 mL Light Blue
13683702 0.5 mL Violet
13683703 1.0 mL Yellow
13683704 2.5 mL Green
13683705 5.0 mL Blue
13683706 10 mL Orange
13683707 25 mL Red
13683708 50 mL Light Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13683700 Supplier Eppendorf™ Supplier No. 0030089405
Edge

Eppendorf's consumable of choice for all Eppendorf Multipette™ multi-dispensers. Available in 9 volume sizes (0.1 to 50 mL), Combitips advanced in 'Eppendorf Quality' dispense in increments as low as 100 mL and are ideally suited for a variety of standard lab applications. Also available in purity grades PCR clean, Biopur™ or Forensic DNA Grade.

Use these tips for comfortable handling while preventing damage to gloves. Eppendorf Combitips advanced™ Standard Pipetter Tips are optimized to meet the increased requirements of a modern laboratory, and offered in three different purity grades to support any application.

  • Combitips advanced are the ideal system components for all Eppendorf Multipettes
  • The 9 volume sizes (0.1 mL – 50 mL) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters depending on the Combitip used
  • Elongated Combitips in volumes 2.5 mL, 5.0 mL and 10 mL make it possible to reach the bottom of the most common laboratory vessels
  • An individual color coding facilitates the quick identification of the desired Combitips

Specifications

Volume (Metric) 0.1 mL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Not Autoclavable
Clean Claims Eppendorf Quality (Non-sterile)
Format Bulk, Bagged
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk. (4 Bags x 25 Pk.)
Product Type Pipette Tips
Color Code White
Disposable Single-use
For Use With (Equipment) Positive Displacement System
Graduated Yes
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less