missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - PCR Clean
GREENER_CHOICE

Catalog No. 13683737
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13683737 0.5 mL
13683736 0.2 mL
13683735 0.1 mL
13683738 1 mL
13683741 10 mL
13683740 5 mL
13683739 2.5 mL
13683743 50 mL
13683742 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13683737 Supplier Eppendorf™ Supplier No. 0030089782
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ensures comfortable handling while preventing damage to gloves. Eppendorf; Combitips advanced™ PCR Pipet Tips meet the increased requirements of a modern laboratory.

Combitips advanced replaces Combitips plus. Functioning according to the positive displacement principle, these always dispense the correct desired volume, independent of the liquid density and its flow characteristics. Working with radioactive, aggressive, toxic or infectious liquids also becomes safer due to hermetically sealed piston preventing aerosol contamination. The new individual color coding facilitates quick identification of the desired tip.

  • Ideal for use with all Eppendorf Repeaters™
  • Nine sizes offer a maximum range of volumes with individual dispensing from 0.1 to 50 mL
  • Funnel geometry prevents damage to gloves and ensures comfortable handling
  • Elongated design allows users to empty all standard laboratory tubes completely
  • Allow for single-hand operation; easy-reach dispenser box conveniently dispenses through a chute
  • Perform automatic Combitip recognition and volume calculation
  • 3D design on the upper rim allows for easy recognition
  • Purity grade: PCR Clean Quality - portioning in 4 resealable bags (25 Tips per bag) ensures dust-free storage
  • Additional purity grades available: standard Eppendorf Quality or Biopur™ (Cat. Nos. 13-683-700 through -726)

Ideal for a range of applications, including ELISA, RIA, PCR set-up and general sample preparation or other serial pipetting tasks.

Specifications

Volume (Metric) 0.5 mL
Sterility Non-sterile
Color Crystal
Autoclavable Not Autoclavable
Clean Claims PCR Clean (Non-sterile)
Format Bulk, Bagged
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk. (4 Reclosable Bags x 25 Pk.)
Product Type Pipet Tips
Color Code Violet
Disposable Single-use
For Use With (Equipment) Eppendorf Multipette
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose PCR Ready
Show More Show Less