Learn More
Eppendorf™ A-4-38 Rotor Adapters
For round and rectangular buckets
$396.71 - $421.58
| Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
|
5702763004
|
Eppendorf™
5702763004 |
9 x 5 mL Tubes |
Pack of 2 Each for $421.58
|
|
|||||
|
5702733008
|
Eppendorf™
5702733008 |
1 x 5 mL Conical Tube |
Pack of 2 Each for $396.71
|
|
|||||
Description
- For use with Centrifuge 5702/ 5702 R/5702 RH and A-4-38 Rotor
- Designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs.
Specifications
| Rotor Adapter | |
| A-4-38 |