missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5810 Series Four- Place Swinging Bucket Tube Adapters

Catalog No. 361007195
Change view
Click to view available options
Bore Size (Metric):
1.1 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bore Size (Metric)
361007195 1.1 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361007195 Supplier Eppendorf™ Supplier No. 022638203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Adapter for the A-4-62 rotor

  • Capacity: 1.2 to 5mL tubes
  • Maximum tube length with or without cap: 115 to 123mm
  • Bore diameter: 11mm

Specifications

For Use With (Equipment) A-4-62 swing bucket rotor for 5810/5810R centrifuge
Bore Size (Metric) 1.1 cm