Learn More
Eppendorf™ 5810 Series Four-Place Swinging Bucket Tube Adapters
Fit in buckets supplied with 05-400-64 swinging bucket rotor to accept tubes from 1.5 to 250mL
$764.37 - $764.37
Specifications
| For Use With (Equipment) | A-4-62 swing bucket rotor for 5810/5810R centrifuge |
|---|
| Catalog Number | Mfr. No. | Bore Size (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Bore Size (Metric) | Price | Quantity | |||||
|
05-400-67
|
Eppendorf™
022638220 |
1.1 cm |
Pack of 2 Each for $764.37
|
|
|||||
|
05-400-68
|
Eppendorf™
022638246 |
1.3 cm |
Pack of 2 Each for $764.37
|
|
|||||
|
05-400-70
|
Eppendorf™
022638289 |
1.75 cm |
Pack of 2 Each for $764.37
|
|
|||||
|
05-400-69
|
Eppendorf™
022638262 |
1.6 cm |
Pack of 2 Each for $764.37
|
|
|||||
|
05-400-78
|
Eppendorf™
022638441 |
6.2 cm |
Pack of 2 Each for $764.37
|
|
|||||
|
05-400-73
|
Eppendorf™
022638343 |
3.1 cm |
Pack of 2 Each for $764.37
|
|
|||||
Description
Adapters are sold in packs of two. Be sure to order the proper number of adapters to fully equip your rotor.
Specifications
| A-4-62 swing bucket rotor for 5810/5810R centrifuge |