missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5810/5810R Series Four-Place Swinging Bucket Rotor

Accommodates 28 x 50mL tubes

Supplier:  Eppendorf™ 022638607

Catalog No. 05412409


Only null left

Description

Description

A-4-81 rotor, for use with Eppendorf Multipurpose Centrifuges 5810/5810R.

  • 4 x 500mL rotor
  • Comes with four swing buckets, each capable of holding 7 x 50mL conical tubes
  • Maximum speed 3,250xg (4,000 rpm)
  • Variety of adapters and capacities available

Specifications

Specifications

Rotor
3,250 x g
5810/5810R Centrifuge
4
4 x 500 mL
4,000 rpm
Four swing bucket
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions