Learn More
Eppendorf™ 5804/5810 Series Tube Adapters for 05-400-81 Fixed Angle Rotor
For use with 05-400-81 Fixed Angle Rotor
Supplier:  Eppendorf™ 022637274
 View more versions of this product
Catalog No. 36-100-7277
Only null left
Explore available promotions
Description
Description
Adapters are supplied in packs of two. Order correct quantity to equip rotor.
Specifications
Specifications
15 mL Conical Tubes
Fixed angle rotor
Product Suggestions
Product Suggestions