Learn More
Eppendorf™ 5804/5810 Series Tube Adapters for 05-400-81 Fixed Angle Rotor
For use with 05-400-81 Fixed Angle Rotor
Supplier:  Eppendorf™ 022637258
 View more versions of this product
Catalog No. 36-100-7275
Only null left
Explore available promotions
Description
Description
Adapters are supplied in packs of two. Order correct quantity to equip rotor.
Specifications
Specifications
50 mL Tubes
Fixed angle rotor
Product Suggestions
Product Suggestions