missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rotor for Centrifuges 5804 R and 5810 R

Eppendorf Rotor for Centrifuges 5804 R and 5810 R-precision meets power. Customize your centrifugation with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-performance applications, ensuring accuracy and reproducibility.

Supplier:  Eppendorf™ 022637401

 View more versions of this product

Catalog No. 0540094


Only null left

Includes: Four tube buckets to accommodate tubes from 1.5 to 100 mL when equipped with accessory adapters

Description

Description

  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.

Order Info

Tube adapters and bucket covers (05-400-109) available separately

Specifications

Specifications

Rotor
4,500 x g
Centrifuges 5804 R and 5810 R
Autoclavable
4 x 100 mL
5,000 rpm
100 mL Rectangular Buckets (x4)
A-4-44
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions