Learn More
Eppendorf™ Rotor for Centrifuge 5418 R
Eppendorf Rotor for Centrifuge 5418 R, precision redefined. Tailor your centrifugation with varied capacities and speeds. Accessorize with adaptors and buckets for versatility in molecular biology and life science applications.
Supplier: Eppendorf™ 022652061
Description
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
- Accessories (adaptors and buckets) further customize your centrifuge setup
Specifications
| FA-45-18-11, Aerosol-tight - Includes rotor lid | |
| 18 x 1.5 to 2.0 mL Tubes | |
| 14,000 rpm | |
| Rotor Lid |
| Aerosol-tight Rotor | |
| 16,873 x g | |
| Centrifuge 5418 R |