missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epmedin C 95.0+%, TCI America™
$132.84 - $392.74
Chemical Identifiers
| CAS | 110642-44-9 |
|---|---|
| Molecular Formula | C39H50O19 |
| Molecular Weight (g/mol) | 822.81 |
| MDL Number | MFCD04039820 |
| InChI Key | ULZLIYVOYYQJRO-JIYCBSMMSA-N |
| Synonym | 3-[[6-Deoxy-2-O-(6-deoxy-alpha-L-mannopyranosyl)-alpha-L-mannopyranosyl]oxy]-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4-oxo-4H-chromen-7-yl beta-D-Glucopyranoside, Baohuoside VI |
| PubChem CID | 5748394 |
| IUPAC Name | 3-[(2S,3R,4R,5R,6S)-4,5-dihydroxy-6-methyl-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one |
| SMILES | CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=C(OC4=C(C3=O)C(=CC(=C4CC=C(C)C)OC5C(C(C(C(O5)CO)O)O)O)O)C6=CC=C(C=C6)OC)C)O)O)O)O)O |
Chemical Identifiers
| 110642-44-9 | |
| 822.81 | |
| ULZLIYVOYYQJRO-JIYCBSMMSA-N | |
| 5748394 | |
| CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=C(OC4=C(C3=O)C(=CC(=C4CC=C(C)C)OC5C(C(C(C(O5)CO)O)O)O)O)C6=CC=C(C=C6)OC)C)O)O)O)O)O |
| C39H50O19 | |
| MFCD04039820 | |
| 3-[[6-Deoxy-2-O-(6-deoxy-alpha-L-mannopyranosyl)-alpha-L-mannopyranosyl]oxy]-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4-oxo-4H-chromen-7-yl beta-D-Glucopyranoside, Baohuoside VI | |
| 3-[(2S,3R,4R,5R,6S)-4,5-dihydroxy-6-methyl-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one |
Specifications
| 110642-44-9 | |
| Yellow | |
| MFCD04039820 | |
| 3-[[6-Deoxy-2-O-(6-deoxy-alpha-L-mannopyranosyl)-alpha-L-mannopyranosyl]oxy]-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4-oxo-4H-chromen-7-yl beta-D-Glucopyranoside, Baohuoside VI | |
| CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=C(OC4=C(C3=O)C(=CC(=C4CC=C(C)C)OC5C(C(C(C(O5)CO)O)O)O)O)C6=CC=C(C=C6)OC)C)O)O)O)O)O | |
| 822.81 | |
| 822.81 | |
| Crystalline Powder |
| 147°C | |
| C39H50O19 | |
| 10 mg | |
| ULZLIYVOYYQJRO-JIYCBSMMSA-N | |
| 3-[(2S,3R,4R,5R,6S)-4,5-dihydroxy-6-methyl-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one | |
| 5748394 | |
| ≥95.0% (HPLC) | |
| Epmedin C |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator