missing translation for 'onlineSavingsMsg'
Learn More

Epmedin C 95.0+%, TCI America™

Catalog No. E132010MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E132010MG 10 mg
E132050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E132010MG Supplier TCI America Supplier No. E132010MG

Chemical Identifiers

CAS 110642-44-9
Molecular Formula C39H50O19
Molecular Weight (g/mol) 822.81
MDL Number MFCD04039820
InChI Key ULZLIYVOYYQJRO-JIYCBSMMSA-N
Synonym 3-[[6-Deoxy-2-O-(6-deoxy-alpha-L-mannopyranosyl)-alpha-L-mannopyranosyl]oxy]-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4-oxo-4H-chromen-7-yl beta-D-Glucopyranoside, Baohuoside VI
PubChem CID 5748394
IUPAC Name 3-[(2S,3R,4R,5R,6S)-4,5-dihydroxy-6-methyl-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OC2C(C(C(OC2OC3=C(OC4=C(C3=O)C(=CC(=C4CC=C(C)C)OC5C(C(C(C(O5)CO)O)O)O)O)C6=CC=C(C=C6)OC)C)O)O)O)O)O

Specifications

Melting Point 147°C
Color Yellow
Quantity 10 mg
Formula Weight 822.81
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Epmedin C
TSCA No
Recommended Storage Refrigerator