missing translation for 'onlineSavingsMsg'
Learn More

Eplerenone, Thermo Scientific Chemicals

Catalog No. p-7214431
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462972500 250 mg
AC462970010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462972500 Supplier Thermo Scientific Chemicals Supplier No. 462972500
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107724-20-9
Molecular Formula C24H30O6
Molecular Weight (g/mol) 414.50
InChI Key JUKPWJGBANNWMW-VWBFHTRKSA-N
IUPAC Name methyl (1'R,2R,2'S,9'R,10'R,11'S,15'S,17'R)-2',15'-dimethyl-5,5'-dioxo-18'-oxaspiro[oxolane-2,14'-pentacyclo[8.8.0.0¹,¹⁷.0²,⁷.0¹¹,¹⁵]octadecan]-6'-ene-9'-carboxylate
SMILES COC(=O)[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@@]23O[C@@H]2C[C@@]2(C)[C@@H](CC[C@@]22CCC(=O)O2)[C@H]13

Specifications

Melting Point 242.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 414.49
Chemical Name or Material Eplerenone

RUO – Research Use Only