missing translation for 'onlineSavingsMsg'
Learn More

Eplerenone 98.0+%, TCI America™

Catalog No. E0905200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0905200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0905200MG Supplier TCI America Supplier No. E0905200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107724-20-9
Molecular Formula C24H30O6
Molecular Weight (g/mol) 417.516
MDL Number MFCD07783647
InChI Key JUKPWJGBANNWMW-NJQGUURESA-N
Synonym 7_,11_,17_-9,11-epoxy-17-hydroxy-3-oxopregn-4-ene-7,21-dicarboxylic acid _-lactone 7-methyl-d3 ester
PubChem CID 122199251
SMILES CC12CCC(=O)C=C1CC(C3C24C(O4)CC5(C3CCC56CCC(=O)O6)C)C(=O)OC

Specifications

Melting Point 242°C
Color White
Quantity 200 mg
Formula Weight 414.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Eplerenone
TSCA No
Recommended Storage Refrigerator