missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Epinephrine (+)-bitartrate salt, 98+%

(-)-Epinephrine (+)-bitartrate salt, CAS # 51-42-3, also known as adrenaline, is a monoamine and catecholamine hormone.

Supplier:  Thermo Scientific Chemicals 430140050

 View more versions of this product

Catalog No. AC430140050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • (-)-Epinephrine (+)-bitartrate salt, is a hormone
  • It is a monoamine and catecholamine

Applications

  • (-)-Epinephrine (+)-bitartrate salt is α-adrenergic and β-adrenergic receptor agonist
  • It can be used in adipose region to minimize blood loss
  • It is potentially a platelet agonist in preparation of optimal platelet aggregometry 96-well plates
Specifications

Specifications

(-)-Epinephrine (+)-bitartrate salt
98.0
155.0°C
0.5% max.
98+%
C13H19NO9
5 g
-20° to -24° (20°C, 589nm) (c=4, 1M HCl)
YLXIPWWIOISBDD-NDAAPVSOSA-N
(2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
124080922
98+%
51-42-3
100.0
White to Yellow
Authentic
Glass bottle
MFCD00035077
15,3674
L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1
333.29
333.29
Fine Crystalline Powder

Chemical Identifiers

51-42-3
333.29
YLXIPWWIOISBDD-NDAAPVSOSA-N
124080922
C13H19NO9
MFCD00035077
L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Fatal if swallowed.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot

GHS Signal Word: Danger

EINECSNumber : 200-097-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only