missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ EpiLife™ CF Kit
GREENER_CHOICE

Catalog No. MEPICF500
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MEPICF500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MEPICF500 Supplier Gibco™ Supplier No. MEPICF500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EpiLife™ CF Kit

A sterile, liquid medium prepared without calcium chloride for the long-term, serum-free culture of human epidermal keratinocytes and human corneal epithelial cells. This basal medium requires the addition of calcium plus an appropriate supplement prior to use. Calcium chloride is provided as a separate component with each bottle of medium. EpiLife greatly extends the in vitro lifespan of human epidermal keratinocytes compared to other serum-free media systems. This basal media is animal origin free and contains essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. It does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. EpiLife™ medium is HEPES and bicarbonate buffered, and is designed for use in an incubator with an atmosphere of 5% CO2 and 95% air.

Specifications

Cell Type Keratinocyte Cells, Epithelial Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type CF Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives HEPES, Phenol Red
Without Additives No Calcium Chloride
Product Line Cascade Biologics, EpiLife
Quantity 500 mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.