missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epigallocatechin Gallate, MP Biomedicals™

Catalog No. MP219916550
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219916550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219916550 Supplier MP Biomedicals Inc Supplier No. 0219916550

Inhibits telomerase and DNA methyltransferase. (-)-Epigallocatechin Gallate, MP Biomedicals is a polyphenolic, tumor-inhibiting constituent of green tea. ECGC is an antioxidant polyphenol flavonoid that inhibits cellular oxidation of low density lipoprote

A natural component of green tea with a variety of biological functions:

  • Bocks the activation of EGF receptors and HER-2 receptors
  • Inhibits fatty acid synthase and glutamate dehydrogenase activity
  • Inhibits the peroxynitrite-induced formation of the oxidative injury markers 8-hydroxy-2-deoxy guanosine and 3-nitrotyrosine, apoptosis in human prostate cancer cells
  • Selectively inhibits activation of JNK/SAPK signaling pathway and suppresses proliferation of vascular smooth muscle cells
  • Shows growth inhibitory effects on tumor cells but not their normal counterparts
  • Inhibits telomerase, a fast-binding and reversible fatty acid synthase inhibitor
  • Inhibits PMA-induced skin thickening
  • Inhibits activation of interleukin-1a mRNA and protein expression
  • Inhibits nitric oxide synthase (iNOS) gene expression
  • At μM concentrations, inhibits VEGF-induced tyrosine phosphorylation

Chemical Identifiers

CAS 989-51-5
Molecular Formula C22H18O11
Molecular Weight (g/mol) 458.375
InChI Key WMBWREPUVVBILR-WIYYLYMNSA-N
Synonym =--epigallocatechin gallate, egcg, epigallocatechin gallate, epigallocatechin 3-gallate, tea catechin, epigallocatechin-3-gallate, =--epigallocatechin-3-o-gallate, teavigo, =--epigallocatechol gallate, epigallocatechin-3-monogallate
PubChem CID 65064
ChEBI CHEBI:4806
IUPAC Name [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O

Specifications

Melting Point 218°C (literature)
Quantity 50 mg
Solubility Information Soluble in water (5mg/mL, clear, faint pink colored solution).
Optical Rotation −185° ± 2° (ethanol) (Lit.)
Formula Weight 458.4g/mol
Percent Purity ≥95%
Physical Form Powder
Recommended Storage Store at 4°C.