missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epigallocatechin Gallate Hydrate 98.0+%, TCI America™

Catalog No. E0694100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0694100MG 100 mg
E0694500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0694100MG Supplier TCI America Supplier No. E0694100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 989-51-5
Molecular Formula C22H18O11
Molecular Weight (g/mol) 458.375
MDL Number MFCD00075940
InChI Key WMBWREPUVVBILR-WIYYLYMNSA-N
Synonym =--epigallocatechin gallate, egcg, epigallocatechin gallate, epigallocatechin 3-gallate, tea catechin, epigallocatechin-3-gallate, =--epigallocatechin-3-o-gallate, teavigo, =--epigallocatechol gallate, epigallocatechin-3-monogallate
PubChem CID 65064
ChEBI CHEBI:4806
IUPAC Name [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O

Specifications

Melting Point 212°C
Quantity 100 mg
Formula Weight 458.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Epigallocatechin Gallate Hydrate
RTECSNumber KB5200000
TSCA No