missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Epidermal Growth Factor Receptor Peptide (985-996)

Catalog No. AAJ64705MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64705MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64705MCR Supplier Thermo Scientific Chemicals Supplier No. J64705MCR
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Content And Storage Store at -20°C
Format Solid
Quantity 1 mg

RUO – Research Use Only