missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin gallate, Thermo Scientific Chemicals

A natural product from green tea that induces apoptosis

Supplier:  Thermo Scientific Chemicals J60814FPL

 View more versions of this product

Catalog No. AAJ60814FPL


Only null left
Explore available promotions

Description

Description

(-)-Epicatechin gallate is used as an antioxidant investigated as an adjunct treatment of some methyicillin resistant bacteria. Epicatechin gallate binds to the cell membrane in methicillin-resistant Staphylococcus aureus which decreases the fluidity of the bilayer and induces changes in gene expression. One of the catechin isomers and a potent antioxidant that can modulate a wide range of membrane proteins. Its bilayer-modifying potency was tested using gramicidin A (gA) channels as probes. All the catechins alter gA channel function and modify bilayer properties, with a 500-fold range in potency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Epicatechin gallate is used as an antioxidant investigated as an adjunct treatment of some methyicillin resistant bacteria. Epicatechin gallate binds to the cell membrane in methicillin-resistant Staphylococcus aureus which decreases the fluidity of the bilayer and induces changes in gene expression. One of the catechin isomers and a potent antioxidant that can modulate a wide range of membrane proteins. Its bilayer-modifying potency was tested using gramicidin A (gA) channels as probes. All the catechins alter gA channel function and modify bilayer properties, with a 500-fold range in potency.

Solubility
Soluble in water, acetone, DMSO, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Specifications

Specifications

(-)-Epicatechin gallate
White
MFCD00075936
=--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate
LSHVYAFMTMFKBA-TZIWHRDSSA-N
[(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
107905
442.37
1257-08-5
C22H18O10
20 mg
Soluble in water,acetone,DMSO,methanol.
C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
442.376
CHEBI:70255
Powder

Chemical Identifiers

1257-08-5
442.376
LSHVYAFMTMFKBA-TZIWHRDSSA-N
107905
[(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
C22H18O10
MFCD00075936
=--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate
CHEBI:70255
C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DH9030000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only