Learn More
Tians epic™ SPP Basic Protection Lab Coats
Sky blue lab coat with white collar, wrists, and snaps is ideal for use in controlled environments and many general application areas.
$170.90 - $170.90
Specifications
| Product Type | Controlled Environments Lab Coat |
|---|---|
| Material | Polypropylene |
| Cuff Style | Knit |
| No. of Pockets | 3 |
| For Use With (Application) | For general protection and safety |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-138-620D
|
Tians
843785XL |
X-Large |
N/A
|
|
|||||
Description
- Latex free, low lint
- For use in controlled environments and many general application areas
- Breathable cloth-like fabric is comfortable and cool
- Features include plastic snaps, knit wrists, knit collar, and three pockets
- Constructed from SPP spunbonded polypropylene fabric
- ISO Cleanroom Class 7
- Neatly folded and cleanroom packaged
Specifications
| Controlled Environments Lab Coat | |
| Knit | |
| For general protection and safety |
| Polypropylene | |
| 3 |