Learn More
Tians Epic™ Premium Coated Barrier Cleanroom Gowns
Static dissipative, lint-free film coated gown for use in critical and low particulate ISO cleanroom class 6 environments.
Supplier: Tians 816851TSXXL
Description
- Latex free, lint-free film coated gown for use in critical and low particulate clean environments
- Static dissipative
- Fabric is suitable for exposure to bloodborne pathogens, some hazardous liquids, and where liquid splash is a concern
- Elastic wrists, elastic thumb loop, wrap-around neck ties, wrap-around waist ties
- Serged seam construction
- Fabric meets or exceeds NIOSH, OSHA, ASHP and ONS guidelines for exposure to chemotherapy drugs
- Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane
- ISO Cleanroom Class 6
- Helmke Drum Testing IEST-RP-CC003.3
Specifications
| Controlled Environments Gown | |
| Neck and Waist Ties | |
| Non-sterile | |
| Static Dissipative |
| Polyethylene, Polypropylene | |
| Single-use | |
| Serged | |
| ASTM F1671, ISO Cleanroom Class 6, IEST-RP-CC003.3 |