Learn More
Tians Epic™ Premium Coated Barrier Cleanroom Gowns
Catalog No. 191303674
Catalog No. 191303674 Supplier Tians Supplier No. 816851TSXL
Static dissipative, lint-free film coated gown for use in critical and low particulate ISO cleanroom class 6 environments.
  • Latex free, lint-free film coated gown for use in critical and low particulate clean environments
  • Static dissipative
  • Fabric is suitable for exposure to bloodborne pathogens, some hazardous liquids, and where liquid splash is a concern
  • Elastic wrists, elastic thumb loop, wrap-around neck ties, wrap-around waist ties
  • Serged seam construction
  • Fabric meets or exceeds NIOSH, OSHA, ASHP and ONS guidelines for exposure to chemotherapy drugs
  • Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane
  • ISO Cleanroom Class 6
  • Helmke Drum Testing IEST-RP-CC003.3
Specifications
Product Type Controlled Environments Gown
Material Polyethylene, Polypropylene
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Seam Type Serged
Resistance Static Dissipative
Certifications/Compliance ASTM F1671, ISO Cleanroom Class 6, IEST-RP-CC003.3